missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl(o-tolyl)phosphine

Catalog No. AAH2750303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2750303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2750303 Supplier Thermo Scientific Chemicals Supplier No. H2750303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5931-53-3 | C19H17P | 276.319 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5931-53-3
Molecular Formula C19H17P
Molecular Weight (g/mol) 276.319
MDL Number MFCD00008515
InChI Key MLBZLJCMHFCTQM-UHFFFAOYSA-N
Synonym diphenyl o-tolyl phosphine, 2-methylphenyl diphenylphosphine, diphenyl-o-tolylphosphine, o-tolyldiphenylphosphine, 2-methylphenyl diphenylphosphane, 2-methylphenyl diphenyl phosphine, diphenyltolylphosphine, acmc-1bwkt, diphenyl 2-methylphenyl phosphine
PubChem CID 80040
IUPAC Name (2-methylphenyl)-diphenylphosphane
SMILES CC1=CC=CC=C1P(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 67°C to 70°C
Quantity 1 g
Formula Weight 276.32
Chemical Name or Material Diphenyl(o-tolyl)phosphine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 227-675-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only