missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl(methyl)sulfonium Tetrafluoroborate 97.0+%, TCI America™

Catalog No. p-7127440
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26851G 1 g
D26855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D26851G Supplier TCI America Supplier No. D26851G

Chemical Identifiers

CAS 10504-60-6
Molecular Formula C13H13BF4S
Molecular Weight (g/mol) 288.11
MDL Number MFCD00051861
InChI Key LHAMVDBAOJCBDW-UHFFFAOYSA-N
Synonym diphenyl methyl sulfonium tetrafluoroborate, methyldiphenylsulphonium tetrafluoroborate 1-, methyldiphenylsulfonium tetrafluoroborate, methyldiphenylsulfanium tetrafluoroborate, acmc-1bx8v, diphenyl methyl sulfoniumtetrafluoroborate, methyl diphenyl sulfanium tetrafluoroborate, diphenyl methyl sulphonium tetrafluoroborate, methyl diphenyl sulfonium tetrafluoroborate
PubChem CID 2758717
IUPAC Name methyldiphenylsulfanium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.C[S+](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 1 g
UN Number 1759
Formula Weight 288.11
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl(methyl)sulfonium Tetrafluoroborate