missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenyl Isophthalate 99.0+%, TCI America™
$134.21 - $268.43
Chemical Identifiers
| CAS | 744-45-6 |
|---|---|
| Molecular Formula | C20H14O4 |
| Molecular Weight (g/mol) | 318.328 |
| MDL Number | MFCD00046051 |
| InChI Key | FHESUNXRPBHDQM-UHFFFAOYSA-N |
| Synonym | diphenyl isophthalate, diphenyl m-phthalate, diphenylisophthalate, unii-wh4w31qij8, 1,3-benzenedicarboxylic acid, diphenyl ester, isophthalic acid diphenyl ester, isophthalic acid, diphenyl ester, wh4w31qij8, 1,3-benzenedicarboxylic acid, 1,3-diphenyl ester, 1,3-diphenyl benzene-1,3-dicarboxylate |
| PubChem CID | 69779 |
| IUPAC Name | diphenyl benzene-1,3-dicarboxylate |
| SMILES | C1=CC=C(C=C1)OC(=O)C2=CC(=CC=C2)C(=O)OC3=CC=CC=C3 |
Chemical Identifiers
| 744-45-6 | |
| 318.328 | |
| FHESUNXRPBHDQM-UHFFFAOYSA-N | |
| 69779 | |
| C1=CC=C(C=C1)OC(=O)C2=CC(=CC=C2)C(=O)OC3=CC=CC=C3 |
| C20H14O4 | |
| MFCD00046051 | |
| diphenyl isophthalate, diphenyl m-phthalate, diphenylisophthalate, unii-wh4w31qij8, 1,3-benzenedicarboxylic acid, diphenyl ester, isophthalic acid diphenyl ester, isophthalic acid, diphenyl ester, wh4w31qij8, 1,3-benzenedicarboxylic acid, 1,3-diphenyl ester, 1,3-diphenyl benzene-1,3-dicarboxylate | |
| diphenyl benzene-1,3-dicarboxylate |
Specifications
| 744-45-6 | |
| White | |
| MFCD00046051 | |
| diphenyl isophthalate, diphenyl m-phthalate, diphenylisophthalate, unii-wh4w31qij8, 1,3-benzenedicarboxylic acid, diphenyl ester, isophthalic acid diphenyl ester, isophthalic acid, diphenyl ester, wh4w31qij8, 1,3-benzenedicarboxylic acid, 1,3-diphenyl ester, 1,3-diphenyl benzene-1,3-dicarboxylate | |
| C1=CC=C(C=C1)OC(=O)C2=CC(=CC=C2)C(=O)OC3=CC=CC=C3 | |
| 318.328 | |
| 318.33 | |
| Crystalline Powder |
| 138°C | |
| C20H14O4 | |
| 10 g | |
| FHESUNXRPBHDQM-UHFFFAOYSA-N | |
| diphenyl benzene-1,3-dicarboxylate | |
| 69779 | |
| ≥99.0% (GC) | |
| Diphenyl Isophthalate |
Safety and Handling
TSCA : Yes