accessibility menu, dialog, popup

UserName

CAS RN 744-45-6

IUPAC Name: 1,3-diphenyl benzene-1,3-dicarboxylate
Synonyms: 1,3-diphenyl benzene-1,3-dicarboxylate
Molecular Weight (g/mol): 318.33
Molecular Formula: C20H14O4
InChi Key: FHESUNXRPBHDQM-UHFFFAOYSA-N
SMILES: O=C(OC1=CC=CC=C1)C1=CC(=CC=C1)C(=O)OC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results