Learn More
Diphenyl ether, 99%
CAS: 101-84-8 | C12H10O | 170.211 g/mol
Supplier: Thermo Scientific Chemicals A157910E
Description
Diphenyl ether is widely used as a heat transfer agent and a dye carrier. It is employed as a processing aid in the production of polyesters. It acts as a chemical intermediate in the production of surface active agents and high temperature lubricants, fire retardants like polybrominated diphenyl ethers (PBDEs) and fragrance for detergents. It plays a vital role in the Ferrario reaction to produce phenoxathiin, which finds application in polyamide and polyimide production.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiphenyl ether is widely used as a heat transfer agent and a dye carrier. It is employed as a processing aid in the production of polyesters. It acts as a chemical intermediate in the production of surface active agents and high temperature lubricants, fire retardants like polybrominated diphenyl ethers (PBDEs) and fragrance for detergents. It plays a vital role in the Ferrario reaction to produce phenoxathiin, which finds application in polyamide and polyimide production.
Solubility
Soluble in alcohol, chloroform, ethyl acetate, benzene, diethyl ether and glacial acetic acid. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Diphenyl ether | |
| 26°C to 29°C | |
| 258°C to 260°C | |
| Aromatic | |
| 1.5805 | |
| 2500 g | |
| 1364620 | |
| diphenyl ether, diphenyl oxide, phenyl ether, oxydibenzene, phenyl oxide, oxybisbenzene, biphenyl oxide, 1,1'-oxydibenzene, benzene, 1,1'-oxybis, oxydiphenyl | |
| USIUVYZYUHIAEV-UHFFFAOYSA-N | |
| phenoxybenzene | |
| 7583 | |
| 170.21 |
| 101-84-8 | |
| 1.071 | |
| 125°C (257°F) | |
| C12H10O | |
| MFCD00003034 | |
| UN3077 | |
| 14,7288 | |
| Soluble in alcohol,chloroform,ethyl acetate,benzene,diethyl ether and glacial acetic acid. Insoluble in water. | |
| C1=CC=C(C=C1)OC2=CC=CC=C2 | |
| 170.211 | |
| CHEBI:39258 | |
| 99% |
Chemical Identifiers
| 101-84-8 | |
| 170.211 | |
| USIUVYZYUHIAEV-UHFFFAOYSA-N | |
| 7583 | |
| phenoxybenzene |
| C12H10O | |
| MFCD00003034 | |
| diphenyl ether, diphenyl oxide, phenyl ether, oxydibenzene, phenyl oxide, oxybisbenzene, biphenyl oxide, 1,1'-oxydibenzene, benzene, 1,1'-oxybis, oxydiphenyl | |
| CHEBI:39258 | |
| C1=CC=C(C=C1)OC2=CC=CC=C2 |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 202-981-2
RTECSNumber : KN8970000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only