missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl ether, 99%

Catalog No. AAA1579136
Change view
Click to view available options
Quantity:
250 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1579136 500 g
AAA1579130 250 g
AAA157910E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1579136 Supplier Thermo Scientific Chemicals Supplier No. A1579136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 101-84-8 | C12H10O | 170.211 g/mol

Diphenyl ether is widely used as a heat transfer agent and a dye carrier. It is employed as a processing aid in the production of polyesters. It acts as a chemical intermediate in the production of surface active agents and high temperature lubricants, fire retardants like polybrominated diphenyl ethers (PBDEs) and fragrance for detergents. It plays a vital role in the Ferrario reaction to produce phenoxathiin, which finds application in polyamide and polyimide production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenyl ether is widely used as a heat transfer agent and a dye carrier. It is employed as a processing aid in the production of polyesters. It acts as a chemical intermediate in the production of surface active agents and high temperature lubricants, fire retardants like polybrominated diphenyl ethers (PBDEs) and fragrance for detergents. It plays a vital role in the Ferrario reaction to produce phenoxathiin, which finds application in polyamide and polyimide production.

Solubility
Soluble in alcohol, chloroform, ethyl acetate, benzene, diethyl ether and glacial acetic acid. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 101-84-8
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.211
MDL Number MFCD00003034
InChI Key USIUVYZYUHIAEV-UHFFFAOYSA-N
Synonym diphenyl ether, diphenyl oxide, phenyl ether, oxydibenzene, phenyl oxide, oxybisbenzene, biphenyl oxide, 1,1'-oxydibenzene, benzene, 1,1'-oxybis, oxydiphenyl
PubChem CID 7583
ChEBI CHEBI:39258
IUPAC Name phenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2

Specifications

Melting Point 26°C to 29°C
Density 1.071
Boiling Point 258°C to 260°C
Flash Point 125°C (257°F)
Odor Aromatic
Refractive Index 1.5805
Quantity 500 g
UN Number UN3077
Beilstein 1364620
Merck Index 14,7288
Solubility Information Soluble in alcohol,chloroform,ethyl acetate,benzene,diethyl ether and glacial acetic acid. Insoluble in water.
Formula Weight 170.21
Percent Purity 99%
Chemical Name or Material Diphenyl ether
Show More Show Less
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 202-981-2
RTECSNumber KN8970000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only