missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenyl Disulfide 99.0+%, TCI America™
Supplier: TCI America P0167100G
Specifications
| Diphenyl Disulfide | |
| 61°C | |
| C12H10S2 | |
| 100 g | |
| diphenyl disulfide, phenyl disulfide, disulfide, diphenyl, diphenyldisulfide, 1,2-diphenyldisulfane, phenyldisulfanyl benzene, diphenyl disulphide, biphenyl disulfide, disulfide diphenyl, phenyldithiobenzene | |
| C1=CC=C(C=C1)SSC2=CC=CC=C2 | |
| 218.332 | |
| 218.33 | |
| Crystalline Powder |
| 882-33-7 | |
| White-Yellow | |
| MFCD00003065 | |
| 3077 | |
| GUUVPOWQJOLRAS-UHFFFAOYSA-N | |
| (phenyldisulfanyl)benzene | |
| 13436 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 882-33-7 | |
| 218.332 | |
| GUUVPOWQJOLRAS-UHFFFAOYSA-N | |
| 13436 | |
| C1=CC=C(C=C1)SSC2=CC=CC=C2 |
| C12H10S2 | |
| MFCD00003065 | |
| diphenyl disulfide, phenyl disulfide, disulfide, diphenyl, diphenyldisulfide, 1,2-diphenyldisulfane, phenyldisulfanyl benzene, diphenyl disulphide, biphenyl disulfide, disulfide diphenyl, phenyldithiobenzene | |
| (phenyldisulfanyl)benzene |
Safety and Handling
EINECSNumber : (3)-1124
RTECSNumber : SS6825000
TSCA : Yes