missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Disulfide 99.0+%, TCI America™

Catalog No. P016725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P016725G 25 g
P0167100G 100 g
P0167500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P016725G Supplier TCI America Supplier No. P016725G

Chemical Identifiers

CAS 882-33-7
Molecular Formula C12H10S2
Molecular Weight (g/mol) 218.332
MDL Number MFCD00003065
InChI Key GUUVPOWQJOLRAS-UHFFFAOYSA-N
Synonym diphenyl disulfide, phenyl disulfide, disulfide, diphenyl, diphenyldisulfide, 1,2-diphenyldisulfane, phenyldisulfanyl benzene, diphenyl disulphide, biphenyl disulfide, disulfide diphenyl, phenyldithiobenzene
PubChem CID 13436
IUPAC Name (phenyldisulfanyl)benzene
SMILES C1=CC=C(C=C1)SSC2=CC=CC=C2

Specifications

Melting Point 61°C
Color White-Yellow
Quantity 25 g
UN Number 3077
Formula Weight 218.33
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl Disulfide
EINECSNumber (3)-1124
RTECSNumber SS6825000
TSCA Yes