Learn More
Diphenyl diselenide, 98%
CAS: 1666-13-3 | C12H10Se2 | 312.15 g/mol
Supplier: Thermo Scientific Chemicals A1391914
Description
Diphenyl diselenide is used in the methoxyselenenylation of alkenes, dihydroxylation of double bonds, hydrothiolation of terminal alkynes. It is used in the synthesis of the unsymmetrical diorganyl selenides, 1-(phenylselenomethyl)vinyl selenides, allylic phenyl selenides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiphenyl diselenide is used in the methoxyselenenylation of alkenes, dihydroxylation of double bonds, hydrothiolation of terminal alkynes. It is used in the synthesis of the unsymmetrical diorganyl selenides, 1-(phenylselenomethyl)vinyl selenides, allylic phenyl selenides.
Solubility
Soluble in water, ether.
Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, halogens, reducing agents.
Specifications
| Diphenyl diselenide | |
| 61°C to 63°C | |
| MFCD00003001 | |
| UN3283 | |
| diphenyl diselenide, diselenide, diphenyl, phenyl diselenide, 1,2-diphenyldiselane, diphenyldiselenide, bis phenylselenide, phenyldiselanyl benzene, diphenyldiselane, ccris 9227, acmc-209duq | |
| YWWZCHLUQSHMCL-UHFFFAOYSA-N | |
| (phenyldiselanyl)benzene | |
| 15460 | |
| 98% |
| 1666-13-3 | |
| C12H10Se2 | |
| 25 g | |
| 2047179 | |
| Soluble in water,ether. | |
| [Se]([Se]C1=CC=CC=C1)C1=CC=CC=C1 | |
| 312.15 | |
| 312.13 |
Chemical Identifiers
| 1666-13-3 | |
| 312.15 | |
| YWWZCHLUQSHMCL-UHFFFAOYSA-N | |
| 15460 | |
| [Se]([Se]C1=CC=CC=C1)C1=CC=CC=C1 |
| C12H10Se2 | |
| MFCD00003001 | |
| diphenyl diselenide, diselenide, diphenyl, phenyl diselenide, 1,2-diphenyldiselane, diphenyldiselenide, bis phenylselenide, phenyldiselanyl benzene, diphenyldiselane, ccris 9227, acmc-209duq | |
| (phenyldiselanyl)benzene |
Safety and Handling
GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P280-P301+P310-P304+P340-P311-P314-P330-P501c
H301+H331-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.
EINECSNumber : 216-780-2
RTECSNumber : JM9152500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only