missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenyl diselenide, 98%
Description
Diphenyl diselenide is used in the methoxyselenenylation of alkenes, dihydroxylation of double bonds, hydrothiolation of terminal alkynes. It is used in the synthesis of the unsymmetrical diorganyl selenides, 1-(phenylselenomethyl)vinyl selenides, allylic phenyl selenides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diphenyl diselenide is used in the methoxyselenenylation of alkenes, dihydroxylation of double bonds, hydrothiolation of terminal alkynes. It is used in the synthesis of the unsymmetrical diorganyl selenides, 1-(phenylselenomethyl)vinyl selenides, allylic phenyl selenides.
Solubility
Soluble in water, ether.
Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, halogens, reducing agents.
Diphenyl diselenide is used in the methoxyselenenylation of alkenes, dihydroxylation of double bonds, hydrothiolation of terminal alkynes. It is used in the synthesis of the unsymmetrical diorganyl selenides, 1-(phenylselenomethyl)vinyl selenides, allylic phenyl selenides.
Solubility
Soluble in water, ether.
Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, halogens, reducing agents.
Specifications
Specifications
| Melting Point | 61°C to 63°C |
| Quantity | 5 g |
| UN Number | UN3283 |
| Beilstein | 2047179 |
| Solubility Information | Soluble in water,ether. |
| Formula Weight | 312.13 |
| Percent Purity | 98% |
| Chemical Name or Material | Diphenyl diselenide |
Safety and Handling
| Hazard Category | H301+H331-H373 |
| Hazard Statement | GHS H Statement H301-H331-H373 Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P301+P310-P304+P340-P311-P314-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S. |
| EINECSNumber | 216-780-2 |
| RTECSNumber | JM9152500 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only