missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Chlorophosphate 95.0+%, TCI America™

Catalog No. D1059500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1059500G 500 g
D105925G 25 g
D1059100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1059500G Supplier TCI America Supplier No. D1059500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2524-64-3
Molecular Formula C12H10ClO3P
Molecular Weight (g/mol) 268.633
MDL Number MFCD00003030
InChI Key BHIIGRBMZRSDRI-UHFFFAOYSA-N
Synonym diphenyl chlorophosphate, diphenyl phosphorochloridate, diphenylchlorophosphate, phosphorochloridic acid, diphenyl ester, chlorodiphenyl phosphate, o,o-diphenyl chlorophosphate, chlorodiphenoxyphosphine oxide, diphenoxychlorophosphine oxide, diphenyl chlorophosphonate, diphenylphosphoric acid monochloride
PubChem CID 75654
IUPAC Name [chloro(phenoxy)phosphoryl]oxybenzene
SMILES C1=CC=C(C=C1)OP(=O)(OC2=CC=CC=C2)Cl

Specifications

Color Yellow
Boiling Point 316°C
Quantity 500 g
UN Number 3265
Formula Weight 268.63
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diphenyl Chlorophosphate
EINECSNumber (3)-3358
TSCA Yes