missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl Carbonate 99.0+%, TCI America™

Catalog No. C004325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C004325G 25 g
C0043500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C004325G Supplier TCI America Supplier No. C004325G

Chemical Identifiers

CAS 102-09-0
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00003037
InChI Key ROORDVPLFPIABK-UHFFFAOYSA-N
Synonym carbonic acid, diphenyl ester, phenyl carbonate, diphenylcarbonate, carbonic acid diphenyl ester, phenyl carbonate pho 2co, unii-ywv401idyn, ph2co3, pho 2co, ywv401idyn, phenyl phenoxyformate
PubChem CID 7597
ChEBI CHEBI:34722
IUPAC Name diphenyl carbonate
SMILES C1=CC=C(C=C1)OC(=O)OC2=CC=CC=C2

Specifications

Melting Point 80°C
Color White
Boiling Point 302°C
Quantity 25 g
UN Number 3077
Formula Weight 214.22
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl Carbonate
EINECSNumber (3)-2490
RTECSNumber FG0500000
TSCA Yes