missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl carbonate, 99%

Catalog No. p-7082092
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1236230 250 g
AAA123620B 1000 g
AAA123620I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1236230 Supplier Thermo Scientific Chemicals Supplier No. A1236230
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 102-09-0 | C13H10O3 | 214.22 g/mol

Used for synthesis of many important organic compounds. As agrochemical intermediates, Dyestuff intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used for synthesis of many important organic compounds. As agrochemical intermediates, Dyestuff intermediate.

Solubility
Soluble in ethanol, diethyl ether, carbon tetrachloride, acetic acid. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 102-09-0
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00003037
InChI Key ROORDVPLFPIABK-UHFFFAOYSA-N
Synonym carbonic acid, diphenyl ester, phenyl carbonate, diphenylcarbonate, carbonic acid diphenyl ester, phenyl carbonate pho 2co, unii-ywv401idyn, ph2co3, pho 2co, ywv401idyn, phenyl phenoxyformate
PubChem CID 7597
ChEBI CHEBI:34722
IUPAC Name diphenyl carbonate
SMILES C1=CC=C(C=C1)OC(=O)OC2=CC=CC=C2

Specifications

Melting Point 76°C to 81°C
Density 1.3
Boiling Point 301°C to 302°C
Flash Point 168°C (334°F)
Odor Mild
Quantity 250 g
Beilstein 1074863
Merck Index 14,3602
Solubility Information Soluble in ethanol,diethyl ether,carbon tetrachloride,acetic acid. Insoluble in water.
Formula Weight 214.22
Percent Purity 99%
Chemical Name or Material Diphenyl carbonate
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 203-005-8
RTECSNumber FG0500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only