missing translation for 'onlineSavingsMsg'
Learn More
Learn More
α,α-Diphenyl-4-piperidinomethanol, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 232520250
| Quantity | 25g |
|---|---|
| Packaging | Glass bottle |
Chemical Identifiers
| C18H21NO | |
| MFCD00066980 | |
| azacyclonol, diphenyl piperidin-4-yl methanol, frenquel, alpha-4-piperidyl benzhydrol, alpha,alpha-diphenyl-4-piperidinomethanol, gamma-pipradol, alpha,alpha-diphenyl-4-piperidinemethanol, ataractan, calmeran, diphenyl-piperidin-4-yl-methanol | |
| diphenyl(piperidin-4-yl)methanol |
Specifications
| α, α-Diphenyl-4-piperidinomethanol | |
| 115-46-8 | |
| 100.0 | |
| Authentic | |
| Glass bottle | |
| 25g | |
| 13, 895 | |
| ZMISODWVFHHWNR-UHFFFAOYSA-N | |
| diphenyl(piperidin-4-yl)methanol | |
| 15723 | |
| 98+% |
| 98+% | |
| 98.0 | |
| 1% max. | |
| 98% min. (HPLC) | |
| C18H21NO | |
| MFCD00066980 | |
| azacyclonol, diphenyl piperidin-4-yl methanol, frenquel, alpha-4-piperidyl benzhydrol, alpha,alpha-diphenyl-4-piperidinomethanol, gamma-pipradol, alpha,alpha-diphenyl-4-piperidinemethanol, ataractan, calmeran, diphenyl-piperidin-4-yl-methanol | |
| C1CNCCC1C(C2=CC=CC=C2)(C3=CC=CC=C3)O | |
| 267.37 | |
| 267.37 |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 204-092-5
RTECSNumber : TN0470000
TSCA : TSCA