missing translation for 'onlineSavingsMsg'
Learn More

α,α-Diphenyl-4-piperidinomethanol, 98+%, Thermo Scientific™

Catalog No. p-122533
Change view
Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC232520050 5g Glass bottle
AC232520250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC232520050 Supplier Thermo Scientific Chemicals Supplier No. 232520050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 115-46-8
Molecular Formula C18H21NO
Molecular Weight (g/mol) 267.37
MDL Number MFCD00066980
InChI Key ZMISODWVFHHWNR-UHFFFAOYSA-N
Synonym azacyclonol, diphenyl piperidin-4-yl methanol, frenquel, alpha-4-piperidyl benzhydrol, alpha,alpha-diphenyl-4-piperidinomethanol, gamma-pipradol, alpha,alpha-diphenyl-4-piperidinemethanol, ataractan, calmeran, diphenyl-piperidin-4-yl-methanol
PubChem CID 15723
IUPAC Name diphenyl(piperidin-4-yl)methanol
SMILES C1CNCCC1C(C2=CC=CC=C2)(C3=CC=CC=C3)O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 5g
Merck Index 13, 895
Formula Weight 267.37
Percent Purity 98+%
Chemical Name or Material α, α-Diphenyl-4-piperidinomethanol, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 204-092-5
RTECSNumber TN0470000
TSCA TSCA

RUO – Research Use Only