missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl-2-pyridylphosphine 97.0+%, TCI America™

Catalog No. p-7127316
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
D24711G 1 g
D24715G 5 g
2 options

Catalog No. D24711G

Supplier: TCI America D24711G

Only null left

Chemical Identifiers

CAS 37943-90-1
Molecular Formula C17H14NP
Molecular Weight (g/mol) 263.28
MDL Number MFCD00192108
InChI Key SVABQOITNJTVNJ-UHFFFAOYSA-N
Synonym diphenyl-2-pyridylphosphine, 2-diphenylphosphino pyridine, 2-pyridyldiphenylphosphine, diphenyl 2-pyridinyl phosphine, diphenyl 2-pyridyl phosphane, 2-diphenylphosphanyl-pyridine, 2-diphenylphosphanyl pyridine, diphenyl pyridin-2-yl phosphane, pph2 2-pyridyl, dpppy
PubChem CID 621893
IUPAC Name diphenyl(pyridin-2-yl)phosphane
SMILES C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=N3

Specifications

Melting Point 85°C
Color White-Yellow
Quantity 1 g
Formula Weight 263.28
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl-2-pyridylphosphine