accessibility menu, dialog, popup

UserName

CAS RN 37943-90-1

IUPAC Name: 2-(diphenylphosphanyl)pyridine
Synonyms: diphenyl-2-pyridylphosphine
Molecular Weight (g/mol): 263.28
Molecular Formula: C17H14NP
InChi Key: SVABQOITNJTVNJ-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=N1
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (4)