missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl-2-pyridylphosphine, 98%

Catalog No. p-7037318
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5602906 5 g
AAH5602914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5602906 Supplier Thermo Scientific Chemicals Supplier No. H5602906
Only null left

CAS: 37943-90-1 | C17H14NP | 263.28 g/mol

It is a ligand for metal-catalyzed carbonylations, hydration, dehydrogenative coupling, carbostannylation, distannylation, and silylation. It is also employed as a reagent for Mitsunobu reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a ligand for metal-catalyzed carbonylations, hydration, dehydrogenative coupling, carbostannylation, distannylation, and silylation. It is also employed as a reagent for Mitsunobu reaction.

Solubility
Soluble in toluene. (almost transparency)

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.

Chemical Identifiers

CAS 37943-90-1
Molecular Formula C17H14NP
Molecular Weight (g/mol) 263.28
MDL Number MFCD00192108
InChI Key SVABQOITNJTVNJ-UHFFFAOYSA-N
Synonym diphenyl-2-pyridylphosphine, 2-diphenylphosphino pyridine, 2-pyridyldiphenylphosphine, diphenyl 2-pyridinyl phosphine, diphenyl 2-pyridyl phosphane, 2-diphenylphosphanyl-pyridine, 2-diphenylphosphanyl pyridine, diphenyl pyridin-2-yl phosphane, pph2 2-pyridyl, dpppy
PubChem CID 621893
IUPAC Name diphenyl(pyridin-2-yl)phosphane
SMILES C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=N3

Specifications

Melting Point 82°C to 84°C
Quantity 5 g
Solubility Information Soluble in toluene. (almost transparency)
Formula Weight 263.27
Percent Purity 98%
Chemical Name or Material Diphenyl-2-pyridylphosphine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only