missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl-2-pyridylphosphine, 97%

Catalog No. AC427660010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427660010 1 g Glass bottle
AC427660050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC427660010 Supplier Thermo Scientific Chemicals Supplier No. 427660010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 37943-90-1 | C17H14NP | 263.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37943-90-1
Molecular Formula C17H14NP
Molecular Weight (g/mol) 263.28
MDL Number MFCD00192108
InChI Key SVABQOITNJTVNJ-UHFFFAOYSA-N
Synonym diphenyl-2-pyridylphosphine, 2-diphenylphosphino pyridine, 2-pyridyldiphenylphosphine, diphenyl 2-pyridinyl phosphine, diphenyl 2-pyridyl phosphane, 2-diphenylphosphanyl-pyridine, 2-diphenylphosphanyl pyridine, diphenyl pyridin-2-yl phosphane, pph2 2-pyridyl, dpppy
PubChem CID 621893
IUPAC Name diphenyl(pyridin-2-yl)phosphane
SMILES C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=N3

Specifications

Melting Point 82.0°C to 84.0°C
Color Tan to White
Elemental Analysis 74.85 to 80.27% C; 5.17 to 5.55% H; 5.1 to 5.6% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 263.28
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl-2-pyridylphosphine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only