missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl (2,3-Dihydro-2-thioxo-3-benzoxazolyl)phosphonate 98.0+%, TCI America™

Catalog No. p-7127079
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20385G 5 g
D203825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20385G Supplier TCI America Supplier No. D20385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111160-56-6
Molecular Formula C19H14NO4PS
Molecular Weight (g/mol) 383.358
MDL Number MFCD00142793
InChI Key POSVXIWJQXSIIP-UHFFFAOYSA-N
Synonym (2,3-Dihydro-2-thioxo-3-benzoxazolyl)phosphonic Acid Diphenyl Ester, DBOP
PubChem CID 11101020
IUPAC Name 3-diphenoxyphosphoryl-1,3-benzoxazole-2-thione
SMILES C1=CC=C(C=C1)OP(=O)(N2C3=CC=CC=C3OC2=S)OC4=CC=CC=C4

Specifications

Melting Point 84°C
Quantity 5 g
Formula Weight 383.36
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl (2,3-Dihydro-2-thioxo-3-benzoxazolyl)phosphonate
EINECSNumber (5)-6555
TSCA Yes