missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenyl-1-pyrenylphosphine 95.0+%, TCI America™
$223.68 - $905.94
Chemical Identifiers
| CAS | 110231-30-6 |
|---|---|
| Molecular Formula | C28H19P |
| Molecular Weight (g/mol) | 386.434 |
| InChI Key | DSYGKYCYNVHCNQ-UHFFFAOYSA-N |
| Synonym | 1-(Diphenylphosphino)pyrene, DPPP |
| PubChem CID | 130626 |
| IUPAC Name | diphenyl(pyren-1-yl)phosphane |
| SMILES | C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C4C=CC5=CC=CC6=C5C4=C(C=C6)C=C3 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D2404100MG
|
TCI America
D2404100MG |
100 mg |
Each for $223.68
|
|
|||||
|
D24041G
|
TCI America
D24041G |
1 g |
Each for $905.94
|
|
|||||
Chemical Identifiers
| 110231-30-6 | |
| 386.434 | |
| 1-(Diphenylphosphino)pyrene, DPPP | |
| diphenyl(pyren-1-yl)phosphane |
| C28H19P | |
| DSYGKYCYNVHCNQ-UHFFFAOYSA-N | |
| 130626 | |
| C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C4C=CC5=CC=CC6=C5C4=C(C=C6)C=C3 |
Specifications
| 110231-30-6 | |
| C28H19P | |
| 1-(Diphenylphosphino)pyrene, DPPP | |
| C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C4C=CC5=CC=CC6=C5C4=C(C=C6)C=C3 | |
| 386.434 | |
| 386.43 | |
| Crystalline Powder |
| 176°C | |
| 100 mg | |
| DSYGKYCYNVHCNQ-UHFFFAOYSA-N | |
| diphenyl(pyren-1-yl)phosphane | |
| 130626 | |
| ≥95.0% (GC) | |
| Diphenyl-1-pyrenylphosphine |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator