missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl-1-pyrenylphosphine 95.0+%, TCI America™

Catalog No. D24041G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24041G 1 g
D2404100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24041G Supplier TCI America Supplier No. D24041G

Chemical Identifiers

CAS 110231-30-6
Molecular Formula C28H19P
Molecular Weight (g/mol) 386.434
InChI Key DSYGKYCYNVHCNQ-UHFFFAOYSA-N
Synonym 1-(Diphenylphosphino)pyrene, DPPP
PubChem CID 130626
IUPAC Name diphenyl(pyren-1-yl)phosphane
SMILES C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C4C=CC5=CC=CC6=C5C4=C(C=C6)C=C3

Specifications

Melting Point 176°C
Quantity 1 g
Formula Weight 386.43
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diphenyl-1-pyrenylphosphine
TSCA Yes
Recommended Storage Refrigerator