missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenic Anhydride 98.0+%, TCI America™
$269.52 - $1291.79
Chemical Identifiers
| CAS | 6050-13-1 |
|---|---|
| Molecular Formula | C14H8O3 |
| Molecular Weight (g/mol) | 224.215 |
| MDL Number | MFCD00004993 |
| InChI Key | RTSGJTANVBJFFJ-UHFFFAOYSA-N |
| Synonym | diphenic anhydride, dibenzo c,e oxepine-5,7-dione, dibenz c,e oxepin-5,7-dione, 2,2'-biphenyldicarboxylic anhydride, 2,2'-diphenyldicarboxylic anhydride, diphenic acid anhydride, 2,2-biphenyldicarboxylic anhydride, dibenz c,7-dione, dibenzo c,e oxepane-5,7-dione |
| PubChem CID | 72794 |
| IUPAC Name | benzo[d][2]benzoxepine-5,7-dione |
| SMILES | C1=CC=C2C(=C1)C3=CC=CC=C3C(=O)OC2=O |
Chemical Identifiers
| 6050-13-1 | |
| 224.215 | |
| RTSGJTANVBJFFJ-UHFFFAOYSA-N | |
| 72794 | |
| C1=CC=C2C(=C1)C3=CC=CC=C3C(=O)OC2=O |
| C14H8O3 | |
| MFCD00004993 | |
| diphenic anhydride, dibenzo c,e oxepine-5,7-dione, dibenz c,e oxepin-5,7-dione, 2,2'-biphenyldicarboxylic anhydride, 2,2'-diphenyldicarboxylic anhydride, diphenic acid anhydride, 2,2-biphenyldicarboxylic anhydride, dibenz c,7-dione, dibenzo c,e oxepane-5,7-dione | |
| benzo[d][2]benzoxepine-5,7-dione |
Specifications
| 6050-13-1 | |
| White-Yellow | |
| MFCD00004993 | |
| diphenic anhydride, dibenzo c,e oxepine-5,7-dione, dibenz c,e oxepin-5,7-dione, 2,2'-biphenyldicarboxylic anhydride, 2,2'-diphenyldicarboxylic anhydride, diphenic acid anhydride, 2,2-biphenyldicarboxylic anhydride, dibenz c,7-dione, dibenzo c,e oxepane-5,7-dione | |
| C1=CC=C2C(=C1)C3=CC=CC=C3C(=O)OC2=O | |
| 224.215 | |
| 224.22 | |
| Crystalline Powder |
| 230°C | |
| C14H8O3 | |
| 5 g | |
| RTSGJTANVBJFFJ-UHFFFAOYSA-N | |
| benzo[d][2]benzoxepine-5,7-dione | |
| 72794 | |
| ≥98.0% (HPLC,T) | |
| Diphenic Anhydride |
Safety and Handling
TSCA : Yes