accessibility menu, dialog, popup

UserName

OOO

CAS RN 6050-13-1

IUPAC Name: 9-oxatricyclo[9.4.0.0²,⁷]pentadeca-1(15),2,4,6,11,13-hexaene-8,10-dione
Synonyms: dibenz c,e oxepin-5,7-dione
Molecular Weight (g/mol): 224.22
Molecular Formula: C14H8O3
InChi Key: RTSGJTANVBJFFJ-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results