missing translation for 'onlineSavingsMsg'
Learn More

Diphenic Anhydride 98.0+%, TCI America™

Catalog No. D08655G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D08655G 5 g
D086525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D08655G Supplier TCI America Supplier No. D08655G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6050-13-1
Molecular Formula C14H8O3
Molecular Weight (g/mol) 224.215
MDL Number MFCD00004993
InChI Key RTSGJTANVBJFFJ-UHFFFAOYSA-N
Synonym diphenic anhydride, dibenzo c,e oxepine-5,7-dione, dibenz c,e oxepin-5,7-dione, 2,2'-biphenyldicarboxylic anhydride, 2,2'-diphenyldicarboxylic anhydride, diphenic acid anhydride, 2,2-biphenyldicarboxylic anhydride, dibenz c,7-dione, dibenzo c,e oxepane-5,7-dione
PubChem CID 72794
IUPAC Name benzo[d][2]benzoxepine-5,7-dione
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C(=O)OC2=O

Specifications

Melting Point 230°C
Color White-Yellow
Quantity 5 g
Formula Weight 224.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Diphenic Anhydride