missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenhydramine 98.0+%, TCI America™
$88.56 - $254.11
Chemical Identifiers
| CAS | 58-73-1 |
|---|---|
| Molecular Formula | C17H21NO |
| Molecular Weight (g/mol) | 255.361 |
| MDL Number | MFCD00274173 |
| InChI Key | ZZVUWRFHKOJYTH-UHFFFAOYSA-N |
| Synonym | 2-Diphenylmethoxy-N,N-dimethylethylamine, 2-(Benzhydryloxy)-N,N-dimethylethylamine |
| PubChem CID | 3100 |
| ChEBI | CHEBI:4636 |
| IUPAC Name | 2-benzhydryloxy-N,N-dimethylethanamine |
| SMILES | CN(C)CCOC(C1=CC=CC=C1)C2=CC=CC=C2 |
Chemical Identifiers
| 58-73-1 | |
| 255.361 | |
| ZZVUWRFHKOJYTH-UHFFFAOYSA-N | |
| 3100 | |
| 2-benzhydryloxy-N,N-dimethylethanamine |
| C17H21NO | |
| MFCD00274173 | |
| 2-Diphenylmethoxy-N,N-dimethylethylamine, 2-(Benzhydryloxy)-N,N-dimethylethylamine | |
| CHEBI:4636 | |
| CN(C)CCOC(C1=CC=CC=C1)C2=CC=CC=C2 |
Specifications
| 58-73-1 | |
| C17H21NO | |
| 5 g | |
| ZZVUWRFHKOJYTH-UHFFFAOYSA-N | |
| 2-benzhydryloxy-N,N-dimethylethanamine | |
| 3100 | |
| 255.36 | |
| Liquid |
| Yellow | |
| MFCD00274173 | |
| 2-Diphenylmethoxy-N,N-dimethylethylamine, 2-(Benzhydryloxy)-N,N-dimethylethylamine | |
| CN(C)CCOC(C1=CC=CC=C1)C2=CC=CC=C2 | |
| 255.361 | |
| CHEBI:4636 | |
| ≥98.0% (GC,T) | |
| Diphenhydramine |
Safety and Handling
EINECSNumber : (9)-0484&(9)-0690
RTECSNumber : KR6825000
TSCA : Yes