missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenhydramine Impurity B Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 4024-34-4 |
| Linear Formula | C18H23 NO · HCl |
| MDL Number | MFCD00550569 |
| Synonym | 2-[(RS)-(4-Methylphenyl)phenylmethoxy]-N,N-dimethylethanamine hydrochloride; 2-[(RS)-(4-METHYLPHENYL) PHENYLMETHOXY]-N,N,DIMETHYLETHANAMINE HYDROCHLORIDE; 2-[(4-Methyl-alpha-phenylbenzyl)oxy]ethyl(dimethyl)ammonium chloride; 4-Methyldiphenhydramine hydrochloride; N,N-Dimethyl-2-[(4-methylphenyl)phenylmethoxy]ethanamin hydrochloride; Neobenodine |
| Type | Certified Reference Material |
| Quantity | 50 mg |
| Formula Weight | 305.84 |
| Grade | Certified Reference Material |