missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Diphenhydramine Hydrochloride USP
Description
Diphenhydramine Hydrochloride USP is a white, odorless, crystalline powder. It slowly darkens on exposure to light.
Specifications
Specifications
| Chemical Name or Material | Diphenhydramine Hydrochloride USP |
| Packaging | Jar, 300mL, White, HDPE, Round, 63-400, TE Pano Neck |
| CAS | 147-24-0 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 291.82 |
| Quantity | 100 g |
| Percent Purity | 98 to 102 % |
| Grade | USP |