missing translation for 'onlineSavingsMsg'
Learn More

Medisca Diphenhydramine Hydrochloride USP

Catalog No. 14185619
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185619 5 g
14185621 100 g
14185387 500 g
14185429 1 kg
14185397 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14185619 Supplier Medisca Supplier No. 028202

Diphenhydramine Hydrochloride USP is a white, odorless, crystalline powder. It slowly darkens on exposure to light.

Diphenhydramine Hydrochloride USP is a white, odorless, crystalline powder. It slowly darkens on exposure to light.

Specifications

Chemical Name or Material Diphenhydramine Hydrochloride USP
Packaging Bottle, 15mL, Amber Glass Wide Mouth Packer, 28-400
CAS 147-24-0
Type Drug Standard
Molecular Weight (g/mol) 291.82
Quantity 5 g
Percent Purity 98 to 102 %
Grade USP