missing translation for 'onlineSavingsMsg'
Learn More

Dipentaerythritol Hexakis(3-mercaptopropionate) 93.0+%, TCI America™

Catalog No. D521225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D521225G 25 g
D5212500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D521225G Supplier TCI America Supplier No. D521225G
Only null left

Chemical Identifiers

CAS 25359-71-1
Molecular Formula C28H46O13S6
Molecular Weight (g/mol) 783.023
MDL Number MFCD00128217
InChI Key YAAUVJUJVBJRSQ-UHFFFAOYSA-N
Synonym DiPETMP
PubChem CID 117451
IUPAC Name [3-(3-sulfanylpropanoyloxy)-2-[[3-(3-sulfanylpropanoyloxy)-2,2-bis(3-sulfanylpropanoyloxymethyl)propoxy]methyl]-2-(3-sulfanylpropanoyloxymethyl)propyl] 3-sulfanylpropanoate
SMILES C(CS)C(=O)OCC(COCC(COC(=O)CCS)(COC(=O)CCS)COC(=O)CCS)(COC(=O)CCS)COC(=O)CCS

Specifications

Color Yellow
Quantity 25 g
Formula Weight 783.02
Percent Purity ≥93.0% (T)
Physical Form Liquid
Chemical Name or Material Dipentaerythritol Hexakis(3-mercaptopropionate)
EINECSNumber (2)-3335
TSCA No