accessibility menu, dialog, popup

UserName

SHOOOOOSHOOHSOOSHOOHSOOSH

CAS RN 25359-71-1

IUPAC Name: 3-{3-[(3-sulfanylpropanoyl)oxy]-2,2-bis({[(3-sulfanylpropanoyl)oxy]methyl})propoxy}-2,2-bis({[(3-sulfanylpropanoyl)oxy]methyl})propyl 3-sulfanylpropanoate
Synonyms: 3-{3-[(3-sulfanylpropanoyl)oxy]-2,2-bis({[(3-sulfanylpropanoyl)oxy]methyl})propoxy}-2,2-bis({[(3-sulfanylpropanoyl)oxy]methyl})propyl 3-sulfanylpropanoate
Molecular Weight (g/mol): 783.02
Molecular Formula: C28H46O13S6
InChi Key: YAAUVJUJVBJRSQ-UHFFFAOYSA-N
SMILES: SCCC(=O)OCC(COCC(COC(=O)CCS)(COC(=O)CCS)COC(=O)CCS)(COC(=O)CCS)COC(=O)CCS
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results