missing translation for 'onlineSavingsMsg'
Learn More

Dioxane, ACS, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor D105020LTMT

 View more versions of this product

Catalog No. 18610262


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dioxane
1
20 L
5e-05
0.0005
Reagent
123-91-1
101°C
Steel Can
0.0016 meq/g
0.99

Chemical Identifiers

123-91-1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.