missing translation for 'onlineSavingsMsg'
Learn More

Dioxane, ACS, 99%, Spectrum™ Chemical

Catalog No. p-7165463
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610261 100 mL Amber Glass Bottle
18610265 500 mL Amber Glass Bottle
18610260 1 L Amber Glass Bottle
18610264 4 L Amber Glass Bottle
18610262 20 L Steel Can
18610263 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18610261 Supplier Spectrum Chemical Mfg Cor Supplier No. D1050100ML
Only null left

Chemical Identifiers

CAS 123-91-1

Specifications

CAS Max % 1
Boiling Point 101°C
Quantity 100 mL
Packaging Amber Glass Bottle
Residue after Evaporation 5e-05
Titratable Acid 0.0016 meq/g
Water 0.0005
Percent Purity 0.99
Grade Reagent
Chemical Name or Material Dioxane

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.