missing translation for 'onlineSavingsMsg'
Learn More

Diosmin, ≥90% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014428
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014428 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014428 Supplier Merck Emd Millipore Supplier No. 6138650MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Diosmin is a flavonoid glycoside that occurs naturally as an active constituent of citrus fruits.(1) It possess a wide range of pharmacological properties like antioxidant, anticarcinogenic, blood-lipid lowering and also anti-inflammatory activities. Diosmin can also find applications in the healing of venous ulcers, enhancement of microcirculation and improvement of venous tone. Featured Industries: Food and Beverages

Specifications

CAS 520-27-4
Physical Form Neat
Linear Formula C28H32O15
MDL Number MFCD00009772
Synonym 3′,5,7-Trihydroxy-4′-methoxyflavone 7-rutinoside; Barosmin; Buchu resin; Diosmetin 7-O-rhamnosylglucoside; Diosmetin 7-rutinoside
Type Analytical Standard
Quantity 50 mg
Formula Weight 608.54
Percent Purity ≥90% (HPLC)
Grade Analytical Standard