Learn More
Thermo Scientific™ Dionex™ Specially Formulated PTFE Tubing
Catalog No. DX160489
Change view
Click to view available options
Description:
250 and 350 um ID
250 um ID; Low pressure
For Use With (Equipment):
Dionex UltiMate 3000 RSLC Nano System
Fused Silica Capillary Tubing
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description For Use With (Equipment)
DX160489 250 and 350 um ID Fused Silica Capillary Tubing
DX160486 250 um ID; Low pressure Dionex UltiMate 3000 RSLC Nano System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DX160489 Supplier Thermo Scientific™ Supplier No. 160489
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Thermo Scientific™ Dionex Specially Formulated PTFE Tubing is used as connector for fused silica tubing (nano and cap LC). Choose fitting dimensions for OD of fused silica tubing used.
Thermo Scientific™ Dionex Specially Formulated PTFE Tubing is used as connector for fused silica tubing (nano and cap LC).
Specifications
Description 250 and 350 um ID
For Use With (Equipment) Fused Silica Capillary Tubing
Product Line nanoLC