Learn More
Thermo Scientific™ Dionex™ PEEK Sleeves
Catalog No. DX161405
Change view
Click to view available options
For Use With (Equipment):
Connections with Microtight Union
Diameter (Metric) Outer:
280 μm
360 μm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Diameter (Metric) Outer
DX161405 Connections with Microtight Union 360 μm
DX161498 Connections with Microtight Union 280 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DX161405 Supplier Thermo Scientific™ Supplier No. 161405
Thermo Scientific™ Dionex PEEK Sleeves are precision cut and polished.
Thermo Scientific™ Dionex PEEK Sleeves are precision cut and polished.
Specifications
Description For microtight union; 360 um OD
For Use With (Equipment) Connections with Microtight Union
Type Tubing Sleeve
Diameter (Metric) Outer 360 μm
Product Line nanoLC