Learn More
Thermo Scientific™ Dionex™ Microtight Fitting
Catalog No. DX161497
Change view
Click to view available options
For Use With (Equipment):
Dionex UltiMate 3000 RSLC Nano System
Catalog No. For Use With (Equipment)
DX161497 Dionex UltiMate 3000 RSLC Nano System
1 options
Catalog No. DX161497 Supplier Thermo Scientific™ Supplier No. 161497
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Thermo Scientific™ Dionex Microtight Fittings are for older generation nano and capillary LC systems. Use fitting sleeve to accommodate required OD of tubing/capillary.
Thermo Scientific™ Dionex Microtight Fittings are for older generation nano and capillary LC systems.
Specifications
Description Microtight fitting
For Use With (Equipment) Dionex UltiMate 3000 RSLC Nano System
Product Line nanoLC