missing translation for 'onlineSavingsMsg'
Learn More

Dioctyl Phthalate, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor D1049200LTBL

 View more versions of this product

Catalog No. 18610257

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

117-81-7
384°C
200 L
Reagent
1
Steel Can
0.98
Liquid

Chemical Identifiers

117-81-7
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.