missing translation for 'onlineSavingsMsg'
Learn More

Dioctyl phthalate-3,4,5,6-d4, MilliporeSigma™ Supelco™

Catalog No. 111018272
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018272 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018272 Supplier Merck Emd Millipore Supplier No. 3419325MG
Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 93952-13-7
Flash Point 109°C - Closed Cup
Physical Form Neat
Linear Formula C6D4 [CO2 CH2(CH2)6 CH3]2
MDL Number MFCD00144150
Melting Point -50°C
Boiling Point 384°C
Type Analytical Standard
Quantity 25 mg
Formula Weight 394.58
Grade Analytical Standard
Show More Show Less