missing translation for 'onlineSavingsMsg'
Learn More

Dioctadecyldimethylammonium Bis(1,3-dithiole-2-thione-4,5-dithiolato)aurate(III), TCI America™

Catalog No. D2134100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2134100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2134100MG Supplier TCI America Supplier No. D2134100MG

Chemical Identifiers

CAS 120141-26-6
Molecular Formula C44H80AuNS10
Molecular Weight (g/mol) 1140.698
MDL Number MFCD00144153
InChI Key DJSOGANPSYSRHM-UHFFFAOYSA-J
Synonym Dimethyldioctadecylammonium Bis(1,3-dithiole-2-thione-4,5-dithiolato)aurate(III)
PubChem CID 44630252
IUPAC Name dimethyl(dioctadecyl)azanium;gold(3+);2-sulfanylidene-1,3-dithiole-4,5-dithiolate
SMILES CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCCCC.C1(=C(SC(=S)S1)[S-])[S-].C1(=C(SC(=S)S1)[S-])[S-].[Au+3]

Specifications

Quantity 100 mg
Formula Weight 1140.70
Physical Form Crystalline Powder
Chemical Name or Material Dioctadecyldimethylammonium Bis(1,3-dithiole-2-thione-4,5-dithiolato)aurate(III)
EINECSNumber (2)-0184&(9)-1971
TSCA No