accessibility menu, dialog, popup

UserName

H3CNH3CCH3CH3SSSSSSSSSSAu

CAS RN 120141-26-6

IUPAC Name: gold(3+) bis((5-sulfanidyl-2-sulfanylidene-2H-1,3-dithiol-4-yl)sulfanide) dimethyldioctadecylazanium
Synonyms: gold(3+) bis((5-sulfanidyl-2-sulfanylidene-1,3-dithiol-4-yl)sulfanide) dimethyldioctadecylazanium
Molecular Weight (g/mol): 1140.7
Molecular Formula: C44H80AuNS10
InChi Key: DJSOGANPSYSRHM-UHFFFAOYSA-J
SMILES: [Au+3].[S-]C1=C([S-])SC(=S)S1.[S-]C1=C([S-])SC(=S)S1.CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCCCC
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results