missing translation for 'onlineSavingsMsg'
Learn More

Dinonyl Phthalate (mixture of isomers) 95.0+%, TCI America™

Catalog No. p030325ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
P030325ML 25 mL
P0303500ML 500 mL
2 options

Catalog No. P030325ML

Supplier: TCI America P030325ML

Only null left

Chemical Identifiers

CAS 84-76-4
Molecular Formula C26H42O4
Molecular Weight (g/mol) 418.62
MDL Number MFCD00036237
InChI Key DROMNWUQASBTFM-UHFFFAOYSA-N
Synonym dinonyl phthalate, dinonylphthalate, nonyl phthalate, di-n-nonyl phthalate, bisoflex dnp, unimoll dn, bisoflex 91, phthalic acid, dinonyl ester, 1,2-benzenedicarboxylic acid, dinonyl ester, bisolflex 91
PubChem CID 6787
IUPAC Name 1,2-dinonyl benzene-1,2-dicarboxylate
SMILES CCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCC

Specifications

Color Yellow
Boiling Point 245°C
Quantity 25 mL
Formula Weight 418.62
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dinonyl Phthalate (mixture of isomers)
EINECSNumber (3)-1307
RTECSNumber TI1800000
TSCA Yes