missing translation for 'onlineSavingsMsg'
Learn More

Dinonyl Phthalate (mixture of isomers) 95.0+%, TCI America™

Catalog No. p-7134546
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P030325ML 25 mL
P0303500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P030325ML Supplier TCI America Supplier No. P030325ML
Only null left

Chemical Identifiers

CAS 84-76-4
Molecular Formula C26H42O4
Molecular Weight (g/mol) 418.62
MDL Number MFCD00036237
InChI Key DROMNWUQASBTFM-UHFFFAOYSA-N
Synonym dinonyl phthalate, dinonylphthalate, nonyl phthalate, di-n-nonyl phthalate, bisoflex dnp, unimoll dn, bisoflex 91, phthalic acid, dinonyl ester, 1,2-benzenedicarboxylic acid, dinonyl ester, bisolflex 91
PubChem CID 6787
IUPAC Name 1,2-dinonyl benzene-1,2-dicarboxylate
SMILES CCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCC

Specifications

Color Yellow
Boiling Point 245°C
Quantity 25 mL
Formula Weight 418.62
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dinonyl Phthalate (mixture of isomers)
EINECSNumber (3)-1307
RTECSNumber TI1800000
TSCA Yes