missing translation for 'onlineSavingsMsg'
Learn More

Dinonyl phthalate, mixture of isomers, 96%

Catalog No. p-7020798
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1287109 10 g
AAA1287118 50 g
AAA1287130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1287109 Supplier Thermo Scientific Chemicals Supplier No. A1287109

CAS: 84-76-4 | C26H42O4 | 418.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 84-76-4
Molecular Formula C26H42O4
Molecular Weight (g/mol) 418.62
MDL Number MFCD00036237
InChI Key DROMNWUQASBTFM-UHFFFAOYSA-N
Synonym dinonyl phthalate, dinonylphthalate, nonyl phthalate, di-n-nonyl phthalate, bisoflex dnp, unimoll dn, bisoflex 91, phthalic acid, dinonyl ester, 1,2-benzenedicarboxylic acid, dinonyl ester, bisolflex 91
PubChem CID 6787
IUPAC Name dinonyl benzene-1,2-dicarboxylate
SMILES CCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCC

Specifications

Density 0.97
Flash Point 216°C (421°F)
Quantity 10 g
Beilstein 1916263
Formula Weight 418.62
Percent Purity 98%
Chemical Name or Material Dinonyl phthalate, mixture of isomers
EINECSNumber 201-560-0
RTECSNumber TI1800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only