missing translation for 'onlineSavingsMsg'
Learn More

Dinonyl phthalate, 96%, mixture of isomers

Catalog No. AC408770250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC408770250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC408770250 Supplier Thermo Scientific Chemicals Supplier No. 408770250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 84-76-4 | C26H42O4 | 418.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 84-76-4
Molecular Formula C26H42O4
Molecular Weight (g/mol) 418.62
MDL Number MFCD00036237
InChI Key DROMNWUQASBTFM-UHFFFAOYSA-N
Synonym dinonyl phthalate, dinonylphthalate, nonyl phthalate, di-n-nonyl phthalate, bisoflex dnp, unimoll dn, bisoflex 91, phthalic acid, dinonyl ester, 1,2-benzenedicarboxylic acid, dinonyl ester, bisolflex 91
PubChem CID 6787
SMILES CCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCC

Specifications

Color Yellow to Brown
Density 0.9700g/mL
Boiling Point 279.0°C to 287.0°C
Flash Point 216°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (sum of isomers) (GC)
Packaging Glass bottle
Refractive Index 1.4825 to 1.4845
Quantity 25 g
Specific Gravity 0.97
Solubility Information Solubility in water: <1g/L (20°)
IUPAC Name 1,2-dinonyl benzene-1,2-dicarboxylate
Viscosity 166 mPa.s (20°C)
Formula Weight 418.61
Percent Purity 96%
Physical Form Oily Liquid
Chemical Name or Material Dinonyl phthalate
Show More Show Less
EINECSNumber 201-560-

RUO – Research Use Only