missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dinitrodurene, 97%, Thermo Scientific™
Spécifications
Spécifications
| Nom ou substance chimique | Dinitrodurene |
| Spectre infrarouge | Authentic |
| Fourchette de pourcentages d’essai | 96% min. (GC) |
| Formule linéaire | (CH3)4C6(NO2)2 |
| Quantité | 5g |
| Beilstein | 05,433 |
| Poids de formule | 224.22 |
| Pourcentage de pureté | 97% |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Very toxic to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing. IF ON SKIN: Gently wash with plenty of soap and water. Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray. Store in a |
| EINECSNumber | 226-766-8 |
RUO – Research Use Only