missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyltin Oxide 95.0+%, TCI America™
Supplier: TCI America D464925G
Specifications
| Dimethyltin Oxide | |
| 395°C | |
| C2H6OSn | |
| 25 g | |
| dimethyltin oxide, stannane, dimethyloxo, dimethyloxostannane, tin, dimethyloxo, dimethyltinoxide, dimethyl oxo tin, dimethyl tin oxide, unii-409qf23wo7, dimethyl oxo stannane, oxodimethyltin | |
| C[Sn](C)=O | |
| 164.78 | |
| 164.78 | |
| Crystalline Powder |
| 2273-45-2 | |
| White | |
| MFCD00013838 | |
| 3146 | |
| WNVQCJNZEDLILP-UHFFFAOYSA-N | |
| dimethylstannanone | |
| 75277 | |
| ≥95.0% (W) |
Chemical Identifiers
| 2273-45-2 | |
| 164.78 | |
| WNVQCJNZEDLILP-UHFFFAOYSA-N | |
| 75277 | |
| C[Sn](C)=O |
| C2H6OSn | |
| MFCD00013838 | |
| dimethyltin oxide, stannane, dimethyloxo, dimethyloxostannane, tin, dimethyloxo, dimethyltinoxide, dimethyl oxo tin, dimethyl tin oxide, unii-409qf23wo7, dimethyl oxo stannane, oxodimethyltin | |
| dimethylstannanone |
Safety and Handling
EINECSNumber : (2)-3013
RTECSNumber : WH7526500
TSCA : Yes