missing translation for 'onlineSavingsMsg'
Learn More

Dimethyltin Oxide 95.0+%, TCI America™

Catalog No. p-7128387
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D464925G 25 g
D4649100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D464925G Supplier TCI America Supplier No. D464925G
Only null left

Chemical Identifiers

CAS 2273-45-2
Molecular Formula C2H6OSn
Molecular Weight (g/mol) 164.78
MDL Number MFCD00013838
InChI Key WNVQCJNZEDLILP-UHFFFAOYSA-N
Synonym dimethyltin oxide, stannane, dimethyloxo, dimethyloxostannane, tin, dimethyloxo, dimethyltinoxide, dimethyl oxo tin, dimethyl tin oxide, unii-409qf23wo7, dimethyl oxo stannane, oxodimethyltin
PubChem CID 75277
IUPAC Name dimethylstannanone
SMILES C[Sn](C)=O

Specifications

Melting Point 395°C
Color White
Quantity 25 g
UN Number 3146
Formula Weight 164.78
Percent Purity ≥95.0% (W)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyltin Oxide
EINECSNumber (2)-3013
RTECSNumber WH7526500
TSCA Yes