missing translation for 'onlineSavingsMsg'
Learn More

Dimethylthiocarbamoyl Chloride 97.0+%, TCI America™

Catalog No. p-7127114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21135G 5 g
D211325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21135G Supplier TCI America Supplier No. D21135G
Only null left

Chemical Identifiers

CAS 16420-13-6
Molecular Formula C3H6ClNS
Molecular Weight (g/mol) 123.60
MDL Number MFCD00004919
InChI Key PHWISQNXPLXQRU-UHFFFAOYSA-N
Synonym dimethylthiocarbamoyl chloride, carbamothioic chloride, dimethyl, chloromethanethioyl dimethylamine, n,n-dimethylthiocarbamoyl chloride, n,n-dimethylcarbamothioic chloride, dimethylthiocarbamoylchloride, carbamoyl chloride, dimethylthio, carbamothioic chloride, n,n-dimethyl, dimethylthiocarbamyl chloride, dimethylthiocarbamic acid chloride
PubChem CID 27871
IUPAC Name (chloromethanethioyl)dimethylamine
SMILES CN(C)C(Cl)=S

Specifications

Melting Point 42°C
Boiling Point 102°C
Quantity 5 g
UN Number 3261
Formula Weight 123.60
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethylthiocarbamoyl Chloride
TSCA Yes
Recommended Storage Refrigerator