accessibility menu, dialog, popup

UserName

CAS RN 16420-13-6

IUPAC Name: (chloromethanethioyl)dimethylamine
Synonyms: (chloromethanethioyl)dimethylamine
Molecular Weight (g/mol): 123.6
Molecular Formula: C3H6ClNS
InChi Key: PHWISQNXPLXQRU-UHFFFAOYSA-N
SMILES: CN(C)C(Cl)=S
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)